ENA428073671 Enamine

1-(2,2-difluoroethoxy)-4-nitro-2-(trifluoromethyl)benzene

  • MDL number MFCD28064043
  • Cas Number : 1803589-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181089
Availability details for
Quantity
  • Availability
  • Add to Cart