ENA428073672 Enamine

4-(isoquinolin-5-yl)piperidin-4-ol dihydrochloride

  • MDL number MFCD28064045
  • Cas Number : 1803608-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181108
Availability details for
Quantity
  • Availability
  • Add to Cart