ENA428073675 Enamine

4-(cyclobutylmethyl)piperidine hydrochloride

  • MDL number MFCD28064048
  • Cas Number : 1803571-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-181151
Availability details for
Quantity
  • Availability
  • Add to Cart