ENA428073680 Enamine

N3-methyl-4-(3-phenyl-1,2,4-oxadiazol-5-yl)-1H-pyrazole-3,5-diamine

  • MDL number MFCD28064053
  • Cas Number : 1188304-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181404
Availability details for
Quantity
  • Availability
  • Add to Cart