ENA428073695 Enamine

1-(3-methylphenyl)-5-(propan-2-yl)-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD28063996
  • Cas Number : 1803571-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-181620
Availability details for
Quantity
  • Availability
  • Add to Cart