ENA428073699 Enamine

5'-bromo-1',2'-dihydrospiro[cyclobutane-1,3'-indole] hydrochloride

  • MDL number MFCD28064002
  • Cas Number : 1803590-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181726
Availability details for
Quantity
  • Availability
  • Add to Cart