ENA428073714 Enamine

2-methyl-5-(pyrrolidin-3-yloxy)pyridine dihydrochloride

  • MDL number MFCD28064063
  • Cas Number : 1803592-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182108
Availability details for
Quantity
  • Availability
  • Add to Cart