ENA428073717 Enamine

N-ethyl-N-[(5-methyl-1,2,4-oxadiazol-3-yl)methyl]azepan-4-amine dihydrochloride

  • MDL number MFCD28064067
  • Cas Number : 1803599-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182145
Availability details for
Quantity
  • Availability
  • Add to Cart