ENA428073718 Enamine

3-(3-ethyl-1,2,4-oxadiazol-5-yl)pyrrolidin-3-ol hydrochloride

  • MDL number MFCD28064069
  • Cas Number : 1803606-50-9
  • Molecular Weight : 219.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-182150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182150
Availability details for
Quantity
  • Availability
  • Add to Cart