ENA428073719 Enamine

2-(3,4-dimethylpiperidin-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28064070
  • Cas Number : 1803590-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182157
Availability details for
Quantity
  • Availability
  • Add to Cart