ENA428073722 Enamine

N-(2-aminoethyl)-N,4-dimethylcyclohexan-1-amine dihydrochloride

  • MDL number MFCD28064073
  • Cas Number : 1803586-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182163
Availability details for
Quantity
  • Availability
  • Add to Cart