ENA428073731 Enamine

2-(3-chloropropyl)-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD28064082
  • Cas Number : 54104-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182395
Availability details for
Quantity
  • Availability
  • Add to Cart