ENA428073732 Enamine

3-[(2-aminoethyl)(methyl)amino]-N-methylpropanamide dihydrochloride

  • MDL number MFCD28064083
  • Cas Number : 1803590-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182396
Availability details for
Quantity
  • Availability
  • Add to Cart