ENA428073733 Enamine

2-[2-(propan-2-yl)pyrrolidin-1-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD28064084
  • Cas Number : 1803560-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-182398
Availability details for
Quantity
  • Availability
  • Add to Cart