ENA428073734 Enamine

(2-aminoethyl)(dicyclopropylmethyl)methylamine dihydrochloride

  • MDL number MFCD28064085
  • Cas Number : 1803606-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182402
Availability details for
Quantity
  • Availability
  • Add to Cart