ENA428073739 Enamine

phenyl 4-oxoazepane-1-carboxylate

  • MDL number MFCD28064090
  • Cas Number : 1798716-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182464
Availability details for
Quantity
  • Availability
  • Add to Cart