ENA428073740 Enamine

3-(1-methylcyclopropyl)-1H-pyrazole hydrochloride

  • MDL number MFCD28064091
  • Cas Number : 1787919-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182475
Availability details for
Quantity
  • Availability
  • Add to Cart