ENA428073746 Enamine

3,4-dihydro-2H-1,4-benzothiazine-6-carbaldehyde hydrochloride

  • MDL number MFCD28064097
  • Cas Number : 1803571-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182518
Availability details for
Quantity
  • Availability
  • Add to Cart