ENA428073747 Enamine

2-amino-N-(1-methyl-1H-pyrazol-3-yl)acetamide dihydrochloride

  • MDL number MFCD28064100
  • Cas Number : 1795465-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182554
Availability details for
Quantity
  • Availability
  • Add to Cart