ENA428073748 Enamine

2-{[(4-bromothiophen-2-yl)methyl]amino}ethan-1-ol hydrochloride

  • MDL number MFCD18912873
  • Cas Number : 1293922-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-179047
Availability details for
Quantity
  • Availability
  • Add to Cart