ENA428073758 Enamine

3-chloro-2-(piperidin-4-yl)pyridine

  • MDL number MFCD05189975
  • Cas Number : 899356-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178528
Availability details for
Quantity
  • Availability
  • Add to Cart