ENA428073762 Enamine

[(5-bromo-2-chlorophenyl)methyl](methyl)amine

  • MDL number MFCD12168572
  • Cas Number : 1096875-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179069
Availability details for
Quantity
  • Availability
  • Add to Cart