ENA428073763 Enamine

6-[(2,2,6,6-tetramethylpiperidin-4-yl)amino]pyridine-3-carbonitrile

  • MDL number MFCD28063930
  • Cas Number : 1389935-19-6
  • Molecular Weight : 258.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-179070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179070
Availability details for
Quantity
  • Availability
  • Add to Cart