ENA428073764 Enamine

potassium 4-(4-fluorophenyl)-2-oxobut-3-enoate

  • Cas Number : 118010-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179072
Availability details for
Quantity
  • Availability
  • Add to Cart