ENA428073768 Enamine

2-(3,4-dimethyl-2,3-dihydro-1H-inden-1-yl)acetic acid

  • MDL number MFCD28023793
  • Cas Number : 1556266-20-6
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-178529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178529
Availability details for
Quantity
  • Availability
  • Add to Cart