ENA428073774 Enamine

3-bromo-5-(propan-2-yloxy)phenol

  • MDL number MFCD12910498
  • Cas Number : 1026796-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179093
Availability details for
Quantity
  • Availability
  • Add to Cart