ENA428073781 Enamine

(3,4,5-triiodophenyl)methanol

  • MDL number MFCD11111446
  • Cas Number : 52273-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-177
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179171
Availability details for
Quantity
  • Availability
  • Add to Cart