ENA428073788 Enamine

(1S)-2-[(9H-fluoren-9-ylmethoxy)carbonyl]-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid

  • MDL number MFCD03788062
  • Cas Number : 204317-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-179184
Availability details for
Quantity
  • Availability
  • Add to Cart