ENA428073791 Enamine

N-(benzyloxy)-4-nitrobenzene-1-sulfonamide

  • MDL number MFCD27991999
  • Cas Number : 1033774-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-178545
Availability details for
Quantity
  • Availability
  • Add to Cart