ENA428073792 Enamine

3-iodo-4-nitroaniline

  • MDL number MFCD15527523
  • Cas Number : 721925-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179195
Availability details for
Quantity
  • Availability
  • Add to Cart