ENA428073798 Enamine

6-fluoro-1H-indole-2-carbaldehyde

  • MDL number MFCD11040760
  • Cas Number : 933746-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178396
Availability details for
Quantity
  • Availability
  • Add to Cart