ENA428073799 Enamine

methyl (2S)-2-methylpyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD22570171
  • Cas Number : 220060-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-131
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179209
Availability details for
Quantity
  • Availability
  • Add to Cart