ENA428073810 Enamine

(2R)-4-aminobutan-2-ol hydrochloride

  • MDL number MFCD26406781
  • Cas Number : 1807941-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178549

Safety Information

msds https://www.enaminestore.com/catalog/EN300-178549
Availability details for
Quantity
  • Availability
  • Add to Cart