ENA428073811 Enamine

3-fluoro-4-[1-methyl-3-(propan-2-yl)-1H-pyrazol-4-yl]benzoic acid

  • MDL number MFCD22268880
  • Cas Number : 1478999-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179226
Availability details for
Quantity
  • Availability
  • Add to Cart