ENA428073815 Enamine

(2S)-4-aminobutan-2-ol hydrochloride

  • MDL number MFCD26406807
  • Cas Number : 1807920-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178550
Availability details for
Quantity
  • Availability
  • Add to Cart