ENA428073821 Enamine

3-methylbut-3-en-1-yl benzoate

  • MDL number MFCD00506908

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179274
Availability details for
Quantity
  • Availability
  • Add to Cart