ENA428073824 Enamine

4-bromo-3-(chloromethyl)benzonitrile

  • MDL number MFCD16037300
  • Cas Number : 853027-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179291
Availability details for
Quantity
  • Availability
  • Add to Cart