ENA428073828 Enamine

2-methylcyclobutan-1-ol

  • MDL number MFCD22054809
  • Cas Number : 1594-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178554
Availability details for
Quantity
  • Availability
  • Add to Cart