ENA428073846 Enamine

6-(dimethylamino)-1-methyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD24081658
  • Cas Number : 1508476-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179339
Availability details for
Quantity
  • Availability
  • Add to Cart