ENA428073848 Enamine

tert-butyl 2-amino-4-(dimethoxymethyl)-1H-imidazole-1-carboxylate

  • MDL number MFCD11976672
  • Cas Number : 917919-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178557
Availability details for
Quantity
  • Availability
  • Add to Cart