ENA428073850 Enamine

methyl 2-amino-5-cyclopropyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD28063726
  • Cas Number : 1692419-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179343
Availability details for
Quantity
  • Availability
  • Add to Cart