ENA428073853 Enamine

3-(propan-2-yl)cyclobutan-1-amine

  • MDL number MFCD19206682
  • Cas Number : 17393-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-178398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178398
Availability details for
Quantity
  • Availability
  • Add to Cart