ENA428073856 Enamine

1-methyl-3-[1-(trifluoromethyl)cyclopropyl]benzene

  • MDL number MFCD28063729
  • Cas Number : 1798713-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-179361
Availability details for
Quantity
  • Availability
  • Add to Cart