ENA428073857 Enamine

ethyl 5-(3-methylphenyl)-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD19482164
  • Cas Number : 1281551-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179362

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179362
Availability details for
Quantity
  • Availability
  • Add to Cart