ENA428073859 Enamine

ethyl 2-(cyclopropylamino)-2-(2,4-dichlorophenyl)acetate hydrochloride

  • MDL number MFCD28063730
  • Cas Number : 1786209-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179365
Availability details for
Quantity
  • Availability
  • Add to Cart