ENA428073867 Enamine

5-methyl-N-(propan-2-yl)-1,2-oxazole-3-carboxamide

  • MDL number MFCD09782758
  • Cas Number : 27144-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-179375
Availability details for
Quantity
  • Availability
  • Add to Cart