ENA428073868 Enamine

ethyl 2-(quinolin-5-yl)acetate

  • MDL number MFCD18416776
  • Cas Number : 1261460-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178399
Availability details for
Quantity
  • Availability
  • Add to Cart