ENA428073870 Enamine

N,N-dipropylpiperazine-1-sulfonamide

  • MDL number MFCD11169491
  • Cas Number : 99868-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178401
Availability details for
Quantity
  • Availability
  • Add to Cart