ENA428073871 Enamine

(4-phenyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD22684343
  • Cas Number : 642462-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179385
Availability details for
Quantity
  • Availability
  • Add to Cart