ENA428073874 Enamine

2-(aminomethyl)-2-[(4-fluorophenyl)methyl]cyclopentan-1-ol hydrochloride

  • MDL number MFCD28063733
  • Cas Number : 1795439-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179408
Availability details for
Quantity
  • Availability
  • Add to Cart